siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019878.4 | NP_063931.1 | 8 | 1822 | ||
| GenBank mRNA | AK033498.1 | - | 8 | 1825 | ||
| AK087595.1 | - | 9 | 1962 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1b1| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019878.4 | NP_063931.1 | 8 | 1822 | ||
| GenBank mRNA | AK033498.1 | - | 8 | 1825 | ||
| AK087595.1 | - | 9 | 1962 | |||