siRNA Details
| Gene Symbol: Ube2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019912.2 | NP_064296.1 | 7 | 1248 | ||
| GenBank mRNA | AK005355.1 | - | 8 | 1197 | ||
| AK136197.1 | - | 7 | 1187 | |||
| AK139623.1 | - | 7 | 1214 | |||
| AK147568.1 | - | 6 | 6934 | |||
| AK148089.1 | - | 7 | 1070 | |||
| AK162093.1 | - | 7 | 1054 | |||
| AK172088.1 | - | 7 | 1075 | |||
| AK207211.1 | - | 1 | 130 | |||
| AK216012.1 | - | 1 | 130 | |||
| AK217849.1 | - | 1 | 125 | |||
| U62483.1 | - | 7 | 1185 | |||

Ube2d2a
Genomic Map