siRNA Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019922.2 | NP_064306.2 | 6 | 1225 | ||
| GenBank mRNA | AJ006469.1 | - | 6 | 1183 | ||
| AK007624.1 | - | 6 | 946 | |||
| AK017797.1 | - | 6 | 1188 | |||
| AK047506.1 | - | 6 | 1181 | |||
| AK191327.1 | - | 2 | 92 | |||
| AK192515.1 | - | 2 | 92 | |||
| AK198132.1 | - | 2 | 92 | |||
| AK199931.1 | - | 2 | 98 | |||
| BC049890.1 | - | 6 | 1162 | 6306169 | ||

Crtap
Genomic Map