siRNA Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019936.3 | NP_064320.1 | 4 | 330 | ||
| GenBank mRNA | AF151637.1 | - | 4 | 295 | ||
| AK004828.1 | - | 4 | 323 | |||
| AK008523.1 | - | 4 | 315 | |||
| AK029409.1 | - | 4 | 318 | |||
| AK213321.1 | - | 2 | 162 | |||
| BC002258.1 | - | 3 | 212 | |||
| BC023013.1 | - | 4 | 304 | 5369938 | ||

Cript
Genomic Map