siRNA Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019936.3 | NP_064320.1 | 5 | 636 | ||
| GenBank mRNA | AF151637.1 | - | 5 | 601 | ||
| AK004828.1 | - | 5 | 630 | |||
| AK008523.1 | - | 5 | 621 | |||
| AK029409.1 | - | 5 | 624 | |||
| BC023013.1 | - | 5 | 610 | 5369938 | ||

Cript
Genomic Map