siRNA Details
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019940.2 | NP_064324.2 | 4 | 2302 | ||
GenBank mRNA | AF167318.1 | - | 2 | 2210 | ||
BC005456.1 | - | 4 | 2238 | 3490047 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019940.2 | NP_064324.2 | 4 | 2302 | ||
GenBank mRNA | AF167318.1 | - | 2 | 2210 | ||
BC005456.1 | - | 4 | 2238 | 3490047 |