siRNA Details
| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 4 | 775 | ||
| GenBank mRNA | AK002800.1 | - | 4 | 776 | ||
| AK005122.1 | - | 4 | 776 | |||
| BC009155.1 | - | 3 | 714 | 2655738 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgst1| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 4 | 775 | ||
| GenBank mRNA | AK002800.1 | - | 4 | 776 | ||
| AK005122.1 | - | 4 | 776 | |||
| BC009155.1 | - | 3 | 714 | 2655738 | ||