siRNA Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164107.1 | NP_001157579.1 | 10 | 1834 | ||
| NM_001164108.1 | NP_001157580.1 | 10 | 1932 | |||
| NM_019955.2 | NP_064339.2 | 10 | 1851 | |||
| GenBank mRNA | AF178953.1 | - | 10 | 1780 | ||
| AK012177.1 | - | 10 | 1824 | |||
| AK154505.1 | - | 10 | 1844 | |||
| BC029210.1 | - | 10 | 1802 | 3590770 | ||
| BF658559.1 | - | 1 | 277 | |||

Ripk3
Genomic Map