siRNA Details
| Gene Symbol: Brinp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019967.2 | NP_064351.2 | 4 | 929 | ||
| GenBank mRNA | AB060589.1 | - | 4 | 929 | ||
| AF202896.1 | - | 3 | 495 | |||
| AK028321.1 | - | 4 | 844 | |||
| AK045420.1 | - | 4 | 811 | |||
| BC048789.1 | - | 4 | 897 | |||
| BC079630.1 | - | 4 | 896 | 5694980 | ||

Brinp1
Genomic Map