siRNA Details
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019971.2 | NP_064355.1 | 4 | 1670 | ||
GenBank mRNA | AF117608.1 | - | 4 | 846 | ||
AF266467.1 | - | 4 | 1670 | |||
AF286725.1 | - | 4 | 649 | |||
AK033734.1 | - | 4 | 1399 | |||
AK042767.1 | - | 4 | 1417 | |||
AK052947.1 | - | 4 | 922 | |||
AK081347.1 | - | 5 | 722 | |||
BC029099.1 | - | 5 | 973 | |||
BC037696.1 | - | 4 | 857 | 4008749 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
