siRNA Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019971.2 | NP_064355.1 | 6 | 2520 | ||
| GenBank mRNA | AF117608.1 | - | 6 | 1696 | ||
| AF266467.1 | - | 6 | 2520 | |||
| AK033734.1 | - | 6 | 2249 | |||
| AK042767.1 | - | 6 | 2267 | |||
| AK052947.1 | - | 6 | 1772 | |||
| BC006027.1 | - | 1 | 568 | |||
| BC029099.1 | - | 7 | 1823 | |||
| BC037696.1 | - | 6 | 1707 | 4008749 | ||

Pdgfc
Genomic Map