siRNA Details
| Gene Symbol: Klk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177373.1 | NP_001170844.1 | 4 | 546 | ||
| NM_019974.2 | NP_064358.1 | 3 | 512 | |||
| GenBank mRNA | AB016226.1 | - | 4 | 396 | ||
| AB016227.1 | - | 4 | 506 | |||
| AK009360.1 | - | 4 | 547 | |||
| AK009659.1 | - | 4 | 521 | |||
| AK009720.1 | - | 4 | 547 | |||
| BC115518.2 | - | 4 | 382 | |||
| BC148310.2 | - | 3 | 221 | |||

Klk11
Genomic Map