siRNA Details
| Gene Symbol: Klk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177373.1 | NP_001170844.1 | 6 | 1095 | ||
| NM_019974.2 | NP_064358.1 | 5 | 1061 | |||
| GenBank mRNA | AB016226.1 | - | 6 | 945 | ||
| AB016227.1 | - | 6 | 1055 | |||
| AK009360.1 | - | 6 | 1096 | |||
| AK009451.1 | - | 2 | 386 | |||
| AK009659.1 | - | 6 | 1070 | |||
| AK009720.1 | - | 6 | 1096 | |||
| AK019102.1 | - | 2 | 386 | |||

Klk11
Genomic Map