siRNA Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019992.3 | NP_064376.1 | 8 | 820 | ||
| GenBank mRNA | AB036058.1 | - | 8 | 810 | ||
| AK041474.1 | - | 8 | 821 | |||
| AK153661.1 | - | 8 | 899 | |||
| AK162655.1 | - | 8 | 902 | |||
| BC145550.1 | - | 7 | 722 | 9054114 | ||
| BC145828.1 | - | 8 | 835 | 40131131 | ||
| BC145830.1 | - | 8 | 836 | 40131133 | ||

Stap1
Genomic Map