siRNA Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 16, 17 | 1408 | ||
| GenBank mRNA | AF139179.1 | - | 16, 17 | 1412 | ||
| BC052702.1 | - | 16, 17 | 1397 | 6400922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Supt20| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 16, 17 | 1408 | ||
| GenBank mRNA | AF139179.1 | - | 16, 17 | 1412 | ||
| BC052702.1 | - | 16, 17 | 1397 | 6400922 | ||