siRNA Details
| Gene Symbol: Clec4n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190320.1 | NP_001177249.1 | 2 | 232 | ||
| NM_020001.2 | NP_064385.1 | 3 | 322 | |||
| GenBank mRNA | AF240357.1 | - | 3 | 291 | ||
| AF240359.1 | - | 3 | 291 | |||
| AK089166.1 | - | 3 | 309 | |||
| AK131697.1 | - | 3 | 309 | |||
| AK154673.1 | - | 2 | 219 | |||
| AK171046.1 | - | 4 | 492 | |||
| BC023008.1 | - | 3 | 291 | 5376247 | ||

Clec4n
Genomic Map