siRNA Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020002.3 | NP_064386.2 | 20 | 2030 | ||
| GenBank mRNA | AF262055.1 | - | 20 | 1960 | ||
| AK077167.1 | - | 20 | 1965 | |||
| AK146832.1 | - | 20 | 2016 | |||
| BC052155.1 | - | 20 | 2026 | 6335959 | ||

Rec8
Genomic Map