siRNA Details
| Gene Symbol: Cdc42ep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163346.1 | NP_001156818.1 | 2 | 2659 | ||
| NM_020006.2 | NP_064390.1 | 2 | 2962 | |||
| GenBank mRNA | AB035088.1 | - | 2 | 3119 | ||
| AK075739.1 | - | 2 | 2652 | |||
| AK146141.1 | - | 2 | 2581 | |||
| AK165318.1 | - | 2 | 2662 | |||
| AK168364.1 | - | 2 | 2581 | |||
| BC003857.1 | - | 2 | 2622 | 3484427 | ||

Cdc42ep4
Genomic Map