siRNA Details
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020032.2 | NP_064416.1 | 7 | 1219 | ||
GenBank mRNA | AF176099.1 | - | 6 | 1160 | ||
AJ131889.1 | - | 7 | 1219 | |||
AK003392.1 | - | 3 | 355 | |||
AK133087.1 | - | 7 | 1188 | |||
BC004767.1 | - | 6 | 1169 | 3156259 |
