siRNA Details
| Gene Symbol: Dhodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020046.3 | NP_064430.1 | 2 | 411 | ||
| GenBank mRNA | AF029667.2 | - | 2 | 116 | ||
| AK013102.1 | - | 2 | 363 | |||
| AK088863.1 | - | 2 | 133 | |||
| AK149350.1 | - | 2 | 159 | |||
| BC019542.1 | - | 2 | 121 | 4039461 | ||
| BC027829.1 | - | 2 | 372 | 5253723 | ||
| BC045206.1 | - | 2 | 361 | 5253764 | ||

Dhodh
Genomic Map