siRNA Details
| Gene Symbol: Dhodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020046.3 | NP_064430.1 | 3 | 616 | ||
| GenBank mRNA | AF029667.2 | - | 3 | 321 | ||
| AK013102.1 | - | 3 | 568 | |||
| AK030798.1 | - | 106 | ||||
| AK088863.1 | - | 3 | 338 | |||
| AK149350.1 | - | 3 | 364 | |||
| BC019542.1 | - | 3 | 326 | 4039461 | ||
| BC027829.1 | - | 3 | 577 | 5253723 | ||
| BC045206.1 | - | 3 | 566 | 5253764 | ||

Dhodh
Genomic Map