siRNA Details
| Gene Symbol: Pik3cg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146200.1 | NP_001139672.1 | 2 | 649 | ||
| NM_001146201.1 | NP_001139673.1 | 2 | 705 | |||
| NM_020272.2 | NP_064668.2 | 2 | 826 | |||
| GenBank mRNA | AF208345.1 | - | 1 | 505 | ||
| AJ249280.1 | - | 2 | 826 | |||
| AK040543.1 | - | 2 | 611 | |||
| AK132553.1 | - | 2 | 651 | |||
| AK137470.1 | - | 2 | 558 | |||
| AK137620.1 | - | 2 | 707 | |||
| BC051246.1 | - | 2 | 566 | 6514979 | ||

Pik3cg
Genomic Map