siRNA Details
| Gene Symbol: Trpm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020277.2 | NP_064673.2 | 7 | 967 | ||
| GenBank mRNA | AB039952.1 | - | 6 | 884 | ||
| AF228681.1 | - | 7 | 967 | |||
| AJ271092.2 | - | 6 | 880 | |||
| AK035197.1 | - | 3 | 497 | |||
| AK044908.1 | - | 3 | 582 | |||
| AY280364.1 | - | 7 | 931 | |||
| AY280365.1 | - | 7 | 931 | |||
| BC133712.1 | - | 6 | 884 | 40143371 | ||

Trpm5
Genomic Map