siRNA Details
| Gene Symbol: Ccl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020279.3 | NP_064675.1 | 3 | 267 | ||
| GenBank mRNA | AF220238.1 | - | 3 | 213 | ||
| AK014821.1 | - | 2 | 1125 | |||
| BC040510.1 | - | 3 | 279 | |||
| BC055864.1 | - | 3 | 270 | 4240191 | ||

Ccl28
Genomic Map