siRNA Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020333.2 | NP_065066.2 | 16 | 2410 | ||
| GenBank mRNA | AF332063.1 | - | 16 | 2080 | ||
| AF332064.1 | - | 16 | 2080 | |||
| AK122460.1 | - | 16 | 2144 | |||
| AK147262.1 | - | 16 | 2412 | |||
| BC054808.1 | - | 16 | 2076 | 6838880 | ||
| KJ535320.1 | - | 16 | 2101 | |||
| KJ535321.1 | - | 16 | 2101 | |||
| KJ535322.1 | - | 16 | 2257 | |||

Slc12a5
Genomic Map