siRNA Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020333.2 | NP_065066.2 | 19 | 2846 | ||
| GenBank mRNA | AF332063.1 | - | 19 | 2516 | ||
| AF332064.1 | - | 19 | 2516 | |||
| AK122460.1 | - | 19 | 2580 | |||
| AK147262.1 | - | 19 | 2848 | |||
| BC054808.1 | - | 19 | 2512 | 6838880 | ||
| KJ535320.1 | - | 19 | 2537 | |||
| KJ535321.1 | - | 19 | 2537 | |||
| KJ535322.1 | - | 19 | 2693 | |||

Slc12a5
Genomic Map