siRNA Details
| Gene Symbol: Sssca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020491.5 | NP_065237.3 | 3 | 283 | ||
| GenBank mRNA | AB012865.1 | - | 3 | 251 | ||
| AK005265.1 | - | 3 | 255 | |||
| AK011957.1 | - | 3 | 273 | |||
| AK012592.1 | - | 3 | 261 | |||
| AK012704.1 | - | 3 | 261 | |||
| BC021593.1 | - | 3 | 253 | 5102521 | ||
| BY081504.1 | - | 3 | 341 | |||

Sssca1
Genomic Map