siRNA Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020506.1 | NP_065252.1 | 3 | 225 | ||
| GenBank mRNA | AF145021.1 | - | 3 | 225 | ||
| AK021022.1 | - | 4 | 856 | |||
| AK084907.1 | - | 4 | 410 | |||
| AK129430.1 | - | 3 | 220 | |||
| AK163516.1 | - | 3 | 506 | |||
| AK163558.1 | - | 4 | 393 | |||
| Gene Symbol: Lats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029242.1 | - | 3112 | |||

Xpo4
Genomic Map