siRNA Details
| Gene Symbol: Ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021299.1 | NP_067274.1 | 5 | 881 | ||
| GenBank mRNA | AB020203.1 | - | 5 | 784 | ||
| AK004864.2 | - | 5 | 896 | |||
| AK008681.1 | - | 5 | 898 | |||
| AK031631.1 | - | 5 | 879 | |||
| AK053644.1 | - | 5 | 941 | |||
| AK089027.1 | - | 5 | 932 | |||
| AK147060.1 | - | 5 | 941 | |||
| AK149996.1 | - | 5 | 938 | |||
| BC016432.1 | - | 5 | 883 | 4217820 | ||
| BC019174.1 | - | 5 | 934 | 5007578 | ||
| BC024871.1 | - | 5 | 881 | 3154594 | ||
| BC058191.1 | - | 5 | 911 | 5340253 | ||

Ak3
Genomic Map