siRNA Details
| Gene Symbol: Lat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020044.3 | NP_064428.1 | 9, 10 | 558 | ||
| NM_022964.4 | NP_075253.2 | 8, 9 | 522 | |||
| GenBank mRNA | AF257136.1 | - | 8, 9 | 502 | ||
| AK134721.1 | - | 6, 7 | 250 | |||
| AK138953.1 | - | 9, 10 | 517 | |||
| AK143533.1 | - | 8, 9 | 523 | |||
| AK144261.1 | - | 9, 10 | 497 | |||
| AK162321.1 | - | 8, 9 | 481 | |||
| AY190024.1 | - | 9, 10 | 523 | |||
| BC005804.1 | - | 8, 9 | 451 | 3154073 | ||

Lat2
Genomic Map