siRNA Details
| Gene Symbol: Tor3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023141.2 | NP_075630.2 | 5 | 887 | ||
| GenBank mRNA | AJ299404.1 | - | 5 | 855 | ||
| AK007814.1 | - | 5 | 876 | |||
| AK009693.1 | - | 5 | 905 | |||
| AK017908.1 | - | 5 | 832 | |||
| AK151048.1 | - | 5 | 873 | |||
| AK151390.1 | - | 6 | 924 | |||
| AK152243.1 | - | 5 | 852 | |||
| AK152358.1 | - | 5 | 888 | |||
| AK152484.1 | - | 5 | 888 | |||
| AK153391.1 | - | 5 | 873 | |||
| AK170296.1 | - | 5 | 882 | |||
| AK172355.1 | - | 5 | 862 | |||
| BC025525.1 | - | 5 | 871 | |||
| BC052851.1 | - | 5 | 1176 | |||
| BC071216.1 | - | 5 | 868 | 30439199 | ||

Tor3a
Genomic Map