siRNA Details
| Gene Symbol: Ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024204.6 | NP_077166.4 | 6 | 1655 | ||
| GenBank mRNA | AK017360.1 | - | 6 | 1656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd22| Gene Symbol: Ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024204.6 | NP_077166.4 | 6 | 1655 | ||
| GenBank mRNA | AK017360.1 | - | 6 | 1656 | ||