siRNA Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015087.1 | - | 5 | 779 | ||
| AK171076.1 | - | 12 | 1797 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_134466, NM_001007150, BC085705, AF483621, AY549447, and others.

Stau2
Genomic Map