siRNA Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025667.3 | NP_079943.2 | 4, 5 | 476 | ||
| GenBank mRNA | AK017503.1 | - | 4, 5 | 472 | ||
| AK079189.1 | - | 4, 5 | 464 | |||
| AK160188.1 | - | 4, 5 | 477 | |||
| BC037609.1 | - | 4, 5 | 437 | 5356647 | ||

Tmem222
Genomic Map