siRNA Details
| Gene Symbol: Odf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162538.1 | NP_001156010.1 | 6 | 716 | ||
| NM_001162539.1 | NP_001156011.1 | 5 | 587 | |||
| NM_025714.4 | NP_079990.2 | 6 | 582 | |||
| GenBank mRNA | AK014630.1 | - | 6 | 584 | ||
| AK016726.1 | - | 6 | 698 | |||
| AK031564.1 | - | 6 | 718 | |||
| AK048931.1 | - | 4 | 2174 | |||
| AK085115.1 | - | 6 | 706 | |||
| BC020075.1 | - | 5 | 556 | 3979963 | ||

Odf2l
Genomic Map