siRNA Details
| Gene Symbol: Odf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162538.1 | NP_001156010.1 | 17 | 2134 | ||
| NM_001162539.1 | NP_001156011.1 | 15 | 1846 | |||
| NM_025714.4 | NP_079990.2 | 16 | 1841 | |||
| GenBank mRNA | AK014630.1 | - | 16 | 1843 | ||
| AK016726.1 | - | 17 | 2116 | |||
| AK220159.1 | - | 6 | 3318 | |||
| BC020075.1 | - | 15 | 1815 | 3979963 | ||

Odf2l
Genomic Map