siRNA Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025918.3 | NP_080194.1 | 5 | 1851 | ||
| GenBank mRNA | AK009319.1 | - | 5 | 1847 | ||
| AK011254.1 | - | 5 | 1852 | |||
| AK028834.1 | - | 5 | 1862 | |||
| AK166820.1 | - | 5 | 1847 | |||
| BC061076.1 | - | 5 | 1860 | 6594359 | ||

Ccdc43
Genomic Map