siRNA Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026504.2 | NP_080780.1 | 1 | 174 | ||
| GenBank mRNA | AK007496.1 | - | 1 | 175 | ||
| AK009825.1 | - | 1 | 170 | |||
| AK014348.1 | - | 1 | 186 | |||
| AK045993.1 | - | 1 | 181 | |||
| AK050334.1 | - | 1 | 181 | |||
| BC052345.1 | - | 1 | 181 | 6585618 | ||

Coq5
Genomic Map