siRNA Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026585.3 | NP_080861.2 | 9 | 916 | ||
| GenBank mRNA | AK038318.1 | - | 9 | 894 | ||
| AK122320.1 | - | 8 | 922 | |||
| AK145716.1 | - | 9 | 917 | |||
| AK161181.1 | - | 9 | 865 | |||
| AK163765.1 | - | 8 | 904 | |||
| AK168388.1 | - | 9 | 903 | |||
| BC032177.1 | - | 7 | 1143 | |||
| BC040393.1 | - | 9 | 840 | |||
| BC049979.1 | - | 9 | 892 | |||
| BC056942.1 | - | 9 | 843 | 6827177 | ||

Fam21
Genomic Map