siRNA Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026585.3 | NP_080861.2 | 19 | 1847 | ||
| GenBank mRNA | AK038318.1 | - | 19 | 1825 | ||
| AK043764.1 | - | 8 | 692 | |||
| AK122320.1 | - | 18 | 1853 | |||
| AK145716.1 | - | 19 | 1848 | |||
| AK161181.1 | - | 19 | 1797 | |||
| AK163765.1 | - | 18 | 1835 | |||
| AK168388.1 | - | 19 | 1834 | |||
| BC032177.1 | - | 17 | 2074 | |||
| BC040393.1 | - | 19 | 1771 | |||
| BC049979.1 | - | 19 | 1823 | |||
| BC056942.1 | - | 19 | 1774 | 6827177 | ||

Fam21
Genomic Map