siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 9 | 900 | ||
| NM_026728.4 | NP_081004.2 | 10 | 968 | |||
| GenBank mRNA | AK005030.1 | - | 10 | 909 | ||
| AK011365.1 | - | 9 | 846 | |||
| AK153878.1 | - | 10 | 943 | |||
| AK160482.1 | - | 8 | 671 | |||
| AK166388.1 | - | 10 | 940 | |||
| AK166965.1 | - | 10 | 910 | |||
| BC025104.1 | - | 10 | 914 | 3591648 | ||
| BC025454.1 | - | 6 | 452 | 5352625 | ||

Echdc2
Genomic Map