siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 9 | 1093 | ||
| NM_026728.4 | NP_081004.2 | 10 | 1161 | |||
| GenBank mRNA | AK005030.1 | - | 10 | 1102 | ||
| AK011365.1 | - | 9 | 1039 | |||
| AK153878.1 | - | 10 | 1136 | |||
| AK160482.1 | - | 8 | 864 | |||
| AK166965.1 | - | 10 | 1103 | |||
| BC025454.1 | - | 6 | 645 | 5352625 | ||

Echdc2
Genomic Map