siRNA Details
| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006808.2 | NP_006799.1 | 4 | 407 | ||
| GenBank mRNA | AJ293399.1 | - | 3 | 272 | ||
| AK223384.1 | - | 4 | 418 | |||
| BC001734.1 | - | 4 | 407 | 3502133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC61B| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006808.2 | NP_006799.1 | 4 | 407 | ||
| GenBank mRNA | AJ293399.1 | - | 3 | 272 | ||
| AK223384.1 | - | 4 | 418 | |||
| BC001734.1 | - | 4 | 407 | 3502133 | ||