siRNA Details
| Gene Symbol: Parp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081009.1 | NP_001074478.1 | 14 | 1610 | ||
| GenBank mRNA | AK149889.1 | - | 13 | 1577 | ||
| AK149918.1 | - | 13 | 1577 | |||
| AK150208.1 | - | 13 | 1577 | |||
| AK150732.1 | - | 12 | 1388 | |||
| AK151510.1 | - | 12 | 1389 | |||
| AK152919.1 | - | 12 | 1388 | |||
| AK153169.1 | - | 12 | 1388 | |||
| AK153270.1 | - | 12 | 1388 | |||
| BC021315.1 | - | 4 | 744 | 5027417 | ||
| BC021881.1 | - | 13 | 1580 | 3710450 | ||
| EF591877.1 | - | 1345 | ||||

Parp8
Genomic Map