siRNA Details
| Gene Symbol: Setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028262.3 | NP_082538.2 | 10 | 1149 | ||
| GenBank mRNA | AK011993.1 | - | 10 | 1129 | ||
| AK031017.1 | - | 2 | 1712 | |||
| AK031371.1 | - | 10 | 1112 | |||
| AK146777.1 | - | 10 | 1143 | |||
| AK166570.1 | - | 10 | 1081 | |||
| AY513271.1 | - | 10 | 1082 | |||
| BC016123.1 | - | 10 | 1079 | 4503842 | ||
| BC019973.1 | - | 10 | 1079 | 4195130 | ||
| BC057968.1 | - | 7 | 769 | 4979330 | ||
| BC108358.1 | - | 10 | 1120 | |||

Setd3
Genomic Map