siRNA Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028731.5 | NP_083007.2 | 15, 16 | 1710 | ||
| GenBank mRNA | AK014813.1 | - | 17, 18 | 1820 | ||
| AK044360.1 | - | 15, 16 | 1712 | |||
| AK157352.1 | - | 15, 16 | 1679 | |||
| BC040392.1 | - | 5, 6 | 393 | |||
| BC052440.1 | - | 5, 6 | 401 | |||
| BC054797.1 | - | 2, 3 | 363 | |||
| BC059230.1 | - | 5, 6 | 401 | |||
| BC138937.1 | - | 15, 16 | 1619 | 8861959 | ||

Esyt2
Genomic Map