siRNA Details
| Gene Symbol: Ppp6r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164159.1 | NP_001157631.1 | 8 | 1015 | ||
| NM_028999.1 | NP_083275.1 | 9 | 1163 | |||
| NM_029456.2 | NP_083732.1 | 9 | 1163 | |||
| GenBank mRNA | AK018652.1 | - | 8 | 1004 | ||
| AK035236.1 | - | 9 | 1143 | |||
| AK088526.1 | - | 8 | 983 | |||
| AK129392.1 | - | 9 | 1105 | |||
| BC008529.1 | - | 9 | 1146 | 3601021 | ||

Ppp6r3
Genomic Map