siRNA Details
| Gene Symbol: Slc52a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029643.3 | NP_083919.1 | 5 | 2435 | ||
| GenBank mRNA | AK029579.1 | - | 5 | 2436 | ||
| AK032015.1 | - | 3 | 2625 | |||
| AK142597.1 | - | 5 | 2517 | |||
More Information
Set Membership: |
|

Slc52a2
Genomic Map