siRNA Details
| Gene Symbol: Nus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030250.2 | NP_084526.1 | 5 | 1082 | ||
| GenBank mRNA | AK155620.1 | - | 5 | 1084 | ||
| AK162260.1 | - | 3 | 244 | |||
| AK167741.1 | - | 5 | 1071 | |||
| AK167939.1 | - | 5 | 1075 | |||
| BC003223.1 | - | 5 | 1041 | 3482163 | ||
| BC018372.1 | - | 5 | 1057 | 3499868 | ||

Nus1
Genomic Map