siRNA Details
| Gene Symbol: D1Ertd622e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133825.3 | NP_598586.1 | 3 | 2106 | ||
| GenBank mRNA | AK135656.1 | - | 3 | 2056 | ||
| AK137265.1 | - | 3 | 2866 | |||
| AK152370.1 | - | 3 | 2078 | |||
| AK168549.1 | - | 3 | 2081 | |||
| BC019375.1 | - | 2 | 1963 | 5008599 | ||
| BC023951.1 | - | 3 | 2075 | 5320263 | ||
| BC034088.1 | - | 3 | 2570 | |||

D1Ertd622e
Genomic Map